missing translation for 'onlineSavingsMsg'
Learn More

Medisca Cyclobenzaprine Hydrochloride, USP

Catalog No. 14185299
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185299 5 g
14185555 500 g
14185553 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14185299 Supplier Medisca Supplier No. 039503

Cyclobenzaprine Hydrochloride USP is a white to off-white, odorless, crystalline powder. It is freely soluble in water, in alcohol, and in methanol.

Cyclobenzaprine Hydrochloride USP is a white to off-white, odorless, crystalline powder. It is freely soluble in water, in alcohol, and in methanol.

Specifications

Chemical Name or Material Cyclobenzaprine Hydrochloride, USP
Packaging Jar 30ml, White, HDPE, Round, 28-400, TRIVENI-DB, CT
CAS 6202-23-9
Type Drug Standard
Molecular Weight (g/mol) 311.85
Quantity 5 g
Percent Purity 98 to 102 %
Grade USP