missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin Y Polyclonal Antibody

Description
Immunogen sequence: GNTTSCCVSS SPKLRRNAHS RLESYRPDTD LSREDTGCNL QHISDRENID DLNMEFNPS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
Specifications
Specifications
| Antigen | Cyclin Y |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CCNY |
| Gene Accession No. | Q8ND76 |
| Gene Alias | 1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; Ccny; CFP1; Cyclin box protein 1; cyclin fold protein 1; cyclin Y; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; cyc-Y; RGD1565969; si:dkey-18n12.2 |
| Gene Symbols | CCNY |
| Show More |