missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin E2 Recombinant Rabbit Monoclonal Antibody (23GB3535)
GREENER_CHOICE

Catalog No. MA553624
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553624 100 μL
MA553625 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553624 Supplier Invitrogen™ Supplier No. MA553624
Only null left

Rabbit Recombinant Monoclonal Antibody

Cyclin E2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.

Specifications

Antigen Cyclin E2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3535
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CCNE2
Gene Accession No. O96020
Gene Alias Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2
Gene Symbols CCNE2
Host Species Rabbit
Immunogen A synthetic peptide of human Cyclin E23 (111-133AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9134
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less