missing translation for 'onlineSavingsMsg'
Learn More

Cyclin E Polyclonal Antibody

Cyclin E Polyclonal Antibody

Supplier:  Thermo Fisher Scientific PA513255

Catalog No. PIPA513255


Only null left
Explore available promotions

Description

Description

Cyclin E Polyclonal Antibody for Western Blot, IHC (P), Flow

Specifications

Specifications

Cyclin E
Polyclonal
Unconjugated
CCNE1
AW538188; Ccne; Ccne1; CycE1; CYCLE; cyclin E; cyclin E1; cyclin Es; cyclin Et; E-S; G1/S-specific cyclin-E1; pCCNE1
Rabbit
Antigen affinity chromatography, Protein A
RUO
898
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P24864
CCNE1
KLH conjugated synthetic peptide between 373-402 amino acids from the C-terminal region of human CCNE1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions