missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin A2 Recombinant Rabbit Monoclonal Antibody (23GB1875)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA554061
Description
Cyclin A2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.Chemical Identifiers
| Cyclin A2 | |
| 1.2 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20248, P51943 | |
| 114494, 12428, 890 | |
| A synthesized peptide derived from human Cyclin A2 (1-250AA). | |
| Antibody |
| 23GB1875 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| CCNA2 | |
| AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg | |
| CCNA2 | |
| Primary | |
| Affinity chromatography |
Specifications
| Cyclin A2 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20248, P51943 | |
| CCNA2 | |
| A synthesized peptide derived from human Cyclin A2 (1-250AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1875 | |
| Unconjugated | |
| CCNA2 | |
| AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114494, 12428, 890 | |
| -20°C | |
| Liquid |