missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin A1/A2 Recombinant Rabbit Monoclonal Antibody (23GB3525)
GREENER_CHOICE

Catalog No. MA553376
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553376 100 μL
MA553377 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553376 Supplier Invitrogen™ Supplier No. MA553376
Only null left

Rabbit Recombinant Monoclonal Antibody

Cyclin A1/A2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The critical role that the family of regulatory proteins known as cyclins play in eukaryotic cell cycle regulation is well established. The best-characterized cyclin complex is the mitotic cyclin B/Cdc2 p34 kinase, the active component of maturing promoting factor. Cyclin A accumulates prior to cyclin B in the cell cycle, appears to be involved in control of S phase and has been shown to associate with cyclin-dependent kinase-2 (Cdk2). In addition, cyclin A has been implicated in cell transformation and is found in complexes with E1A, transcription factors DRTF1 and E2F and retinoblastoma protein, p110. A second form of cyclin A, named cyclin A1 because of its high sequence homology to Xenopus cyclin A1, is most highly expressed in germ cells. It has been proposed that cyclin A1 can associate with Cdk2, p39 and Cdc2 p34.

Specifications

Antigen Cyclin A1/A2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3525
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CCNA2
Gene Accession No. P20248, P51943, P78396, Q61456, Q6AY13
Gene Alias AA408589; CCN1; Ccn-1; CCNA; Ccna1; CCNA2; CT146; Cyca; Cyca2; Cyclin A; cyclin A1; cyclin A2; Cyclin A2 (Cyclin A); CyclinA; cyclin-A; CyclinA1; Cyclin-A1; cyclin-A2; testicular tissue protein Li 34
Gene Symbols CCNA1, CCNA2
Host Species Rabbit
Immunogen A synthetic peptide of human Cyclin A1/A2 (250-465AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114494, 12427, 12428, 295052, 890, 8900
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less