missing translation for 'onlineSavingsMsg'
Learn More

Cyclic Pifithrin-alpha Hydrobromide 98.0+%, TCI America™

Catalog No. C2826100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2826100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2826100MG Supplier TCI America Supplier No. C2826100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 511296-88-1
Molecular Formula C16H17BrN2S
Molecular Weight (g/mol) 349.29
MDL Number MFCD02683960
InChI Key SGNCOAOESGSEOP-UHFFFAOYSA-N
Synonym pifithrin-beta hydrobromide, cyclic pifithrin-alpha hydrobromide, pifithrin-alpha, cyclic, unii-up003xxt4d, up003xxt4d, 2-4-methylphenyl imidazo 2,1-b-5,6,7,8-tetrahydrobenzothiazole, hbr, 2-4-methylphenyl imidazo 2,1-b-5,6,7,8-tetrahydrobenzothiazole hydrobromide, cyclic pifithrin-, a hydrobromide, pifithrin-beta hydrobromide mi, cyclic-pifithrin-alpha
PubChem CID 11515812
IUPAC Name 4-(4-methylphenyl)-7-thia-2,5-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),3,5-triene hydrobromide
SMILES Br.CC1=CC=C(C=C1)C1=CN2C(SC3=C2CCCC3)=N1

Specifications

Color White
Quantity 100 mg
Formula Weight 349.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclic Pifithrin-alpha Hydrobromide
TSCA No
Recommended Storage Freezer