accessibility menu, dialog, popup

UserName

H3CNSNHBr

CAS RN 511296-88-1

IUPAC Name: 4-(4-methylphenyl)-7-thia-2,5-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),3,5-triene hydrobromide
Synonyms: 4-(4-methylphenyl)-7-thia-2,5-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),3,5-triene hydrobromide
Molecular Weight (g/mol): 349.29
Molecular Formula: C16H17BrN2S
InChi Key: SGNCOAOESGSEOP-UHFFFAOYSA-N
SMILES: Br.CC1=CC=C(C=C1)C1=CN2C(SC3=C2CCCC3)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results