missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYBA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599812
Description
Antibody detects endogenous levels of total CYBA.
Cytochrome b is comprised of a light chain (alpha) and a heavy chain (beta). This gene encodes the light, alpha subunit which has been proposed as a primary component of the microbicidal oxidase system of phagocytes. Mutations in this gene are associated with autosomal recessive chronic granulomatous disease (CGD), that is characterized by the failure of activated phagocytes to generate superoxide, which is important for the microbicidal activity of these cells.Specifications
| CYBA | |
| Polyclonal | |
| Unconjugated | |
| CYBA | |
| b558; CYBA; cytochrome b light chain; cytochrome b(558) alpha chain; cytochrome b(558) alpha-subunit; cytochrome b, alpha polypeptide; cytochrome b-245 alpha chain; cytochrome b-245 light chain; cytochrome b-245, alpha polypeptide; cytochrome b558 alpha-subunit; cytochrome b558 subunit alpha; cytochrome beta-558; flavocytochrome b-558 alpha polypeptide; neutrophil cytochrome b 22 kDa polypeptide; nmf333; p22 phagocyte B-cytochrome; p22 phox; p22phox; p22-PHOX; superoxide-generating NADPH oxidase light chain subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13057, 1535, 79129 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13498, Q61462, Q62737 | |
| CYBA | |
| A synthesized peptide derived from human CYBA(Accession P13498), corresponding to amino acid residues M1-R32. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |