missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric fluoride, 98%

Catalog No. p-7022760
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1566606 5 g
AAA1566609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1566606 Supplier Thermo Scientific Chemicals Supplier No. A1566606
Only null left

CAS: 675-14-9 | C3F3N3 | 135.049 g/mol

Cyanuric fluoride acts as a fluorinating agent used in the conversion of carboxylic acids into acyl fluorides. It is used as a precursor for fibre-reactive dyes. It is a specific reagent for tyrosine residues in enzyme. Further, it is involved in the preparation of cyanuric acid by hydrolysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanuric fluoride acts as a fluorinating agent used in the conversion of carboxylic acids into acyl fluorides. It is used as a precursor for fibre-reactive dyes. It is a specific reagent for tyrosine residues in enzyme. Further, it is involved in the preparation of cyanuric acid by hydrolysis

Solubility
Miscible with chloroform, carbon tetrachloride,ether, dioxane and ketones.

Notes
May precipitate upon storage. Moisture sensitive. Incompatible strong oxidizing agents, strong bases and alcohols.

Chemical Identifiers

CAS 675-14-9
Molecular Formula C3F3N3
MDL Number MFCD00014597
InChI Key VMKJWLXVLHBJNK-UHFFFAOYSA-N
Synonym cyanuric fluoride, trifluorotriazine, cyanuric trifluoride, trifluoro-s-triazine, 1,3,5-triazine, 2,4,6-trifluoro, 2,4,6-trifluoro-s-triazine, s-triazine, 2,4,6-trifluoro, cyanuryl fluoride, unii-31kw4s5fbb, s-triazine, 2,4,6-trifluro
PubChem CID 12664

Specifications

Melting Point -38°C
Density 1.574
Boiling Point 73°C to 74°C
Odor Acrid
Refractive Index 1.384
Quantity 5 g
UN Number UN3389
Beilstein 124237
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Miscible with chloroform,carbon tetrachloride,ether,dioxane and ketones.
SMILES C1(=NC(=NC(=N1)F)F)F
IUPAC Name 2,4,6-trifluoro-1,3,5-triazine
Molecular Weight (g/mol) 135.049
Formula Weight 135.05
Percent Purity 98%
Chemical Name or Material Cyanuric fluoride
Show More Show Less
Hazard Category H310+H330-H314-H335
Hazard Statement GHS H Statement
H310-H330-H314-H318
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.
EINECSNumber 211-620-8
RTECSNumber XZ1750000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only