missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric Chloride 98.0+%, TCI America™

Catalog No. C0460500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0460500G 500 g
C046025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0460500G Supplier TCI America Supplier No. C0460500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.40
MDL Number MFCD00006046
InChI Key MGNCLNQXLYJVJD-UHFFFAOYSA-N
Synonym cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
PubChem CID 7954
ChEBI CHEBI:58964
IUPAC Name trichloro-1,3,5-triazine
SMILES ClC1=NC(Cl)=NC(Cl)=N1

Specifications

Melting Point 147°C
Color White
Quantity 500 g
UN Number 2670
Formula Weight 184.4
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyanuric Chloride
EINECSNumber (5)-1045
RTECSNumber XZ1400000
TSCA Yes