accessibility menu, dialog, popup

UserName

ClNClNClN

CAS RN 108-77-0

IUPAC Name: trichloro-1,3,5-triazine
Synonyms: cyanuric chloride|cyanuryl chloride|cyanuric acid chloride|cyanuric trichloride|s-triazine trichloride|tricyanogen chloride|trichlorocyanidine|cyanuric acid trichloride|sym-trichlorotriazine
Molecular Weight (g/mol): 184.4
Molecular Formula: C3Cl3N3
InChi Key: MGNCLNQXLYJVJD-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=NC(Cl)=N1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)