missing translation for 'onlineSavingsMsg'
Learn More

(Cyanomethyl)triphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. C1739100G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1739100G 100 g
C173925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1739100G Supplier TCI America Supplier No. C1739100G

Chemical Identifiers

CAS 4336-70-3
Molecular Formula C20H17ClNP
Molecular Weight (g/mol) 337.79
MDL Number MFCD00031672
InChI Key ARPLQAMUUDIHIT-UHFFFAOYSA-M
Synonym cyanomethyl triphenylphosphonium chloride, phosphonium, cyanomethyl triphenyl-, chloride, cyanomethyl triphenylphosphanium chloride, cyanomethyl triphenyl phosphanium chloride, cyanomethyltriphenylphosphonium chloride, acmc-1ctgb, wln: nc1pr&r&r &g, cyanomethyl triphenylphosphoniumchloride, cyanomethyl triphenyl phosphonium chloride
PubChem CID 197040
IUPAC Name (cyanomethyl)triphenylphosphanium chloride
SMILES [Cl-].N#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 100 g
UN Number 3439
Formula Weight 337.79
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Cyanomethyl)triphenylphosphonium Chloride
RTECSNumber TA2075000
TSCA No