accessibility menu, dialog, popup

UserName

ClNP

CAS RN 4336-70-3

IUPAC Name: (cyanomethyl)triphenylphosphanium chloride
Synonyms: (cyanomethyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 337.79
Molecular Formula: C20H17ClNP
InChi Key: ARPLQAMUUDIHIT-UHFFFAOYSA-M
SMILES: [Cl-].N#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)