missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanomethyl Formate 98.0+%, TCI America™
Supplier: TCI America F035625G
Specifications
| Cyanomethyl Formate [Formylating Reagent] | |
| Colorless | |
| C3H3NO2 | |
| 25 g | |
| ZFLAEHBSVFWEHW-UHFFFAOYSA-N | |
| cyanomethyl formate | |
| 4205685 | |
| ≥98.0% (GC) |
| 150760-95-5 | |
| 64°C | |
| MFCD00661077 | |
| Formic Acid Cyanomethyl Ester | |
| O=COCC#N | |
| 85.06 | |
| 85.06 | |
| Liquid |
Chemical Identifiers
| 150760-95-5 | |
| 85.06 | |
| ZFLAEHBSVFWEHW-UHFFFAOYSA-N | |
| 4205685 | |
| O=COCC#N |
| C3H3NO2 | |
| MFCD00661077 | |
| Formic Acid Cyanomethyl Ester | |
| cyanomethyl formate |
Safety and Handling
TSCA : No