accessibility menu, dialog, popup

UserName

NOO

CAS RN 150760-95-5

IUPAC Name: cyanomethyl formate
Synonyms: cyanomethyl formate
Molecular Weight (g/mol): 85.06
Molecular Formula: C3H3NO2
InChi Key: ZFLAEHBSVFWEHW-UHFFFAOYSA-N
SMILES: O=COCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results