missing translation for 'onlineSavingsMsg'
Learn More

Cyanomethyl Formate 98.0+%, TCI America™

Catalog No. F03565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03565G 5 g
F035625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03565G Supplier TCI America Supplier No. F03565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150760-95-5
Molecular Formula C3H3NO2
Molecular Weight (g/mol) 85.06
MDL Number MFCD00661077
InChI Key ZFLAEHBSVFWEHW-UHFFFAOYSA-N
Synonym Formic Acid Cyanomethyl Ester
PubChem CID 4205685
IUPAC Name cyanomethyl formate
SMILES O=COCC#N

Specifications

Color Colorless
Boiling Point 64°C
Quantity 5 g
Formula Weight 85.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyanomethyl Formate [Formylating Reagent]