missing translation for 'onlineSavingsMsg'
Learn More

Cyanocobalamin, Crystalline, USP, 96-102%, Spectrum™ Chemical

Catalog No. 18601934
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601934 5 g Amber Glass Bottle
18601935 25 g Amber Glass Bottle
18601936 100 g Amber Glass Bottle
18601937 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601934 Supplier Spectrum Chemical Mfg Cor Supplier No. CY1055GM
Get Up to $500 in Chemicals with Purchase
View all promotions

CY105, 68-19-9, C63H88CoN14O14P

Spectrum™ Chemical Cyanocobalamin, Crystalline, USP is a man-made form of vitamin B12 used to treat low levels (deficiency) of this vitamin. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 68-19-9
Molecular Formula C63H88CoN14O14P
Molecular Weight (g/mol) 1355.39
MDL Number MFCD00151092
InChI Key YUWVGNQGDAPKCX-BVWPOUIRNA-L
IUPAC Name (10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ5,9,19,26λ5,43,44λ5,45λ5-heptaaza-15λ5-phospha-1-cobaltadodecacyclo[27.14.1.11,34.12,9.110,13.01,26.03,8.023,27.025,42.032,44.039,43.037,45]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
SMILES [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O

Specifications

CAS Max % 100%
Loss on Drying 12%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 96 to 102%
Grade USP