missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanoacetohydrazide 97.0+%, TCI America™
Supplier: TCI America C080025G
Specifications
| Cyanoacetohydrazide | |
| 107°C | |
| MFCD00007611 | |
| 2811 | |
| HPHBOJANXDKUQD-UHFFFAOYSA-N | |
| 2-cyanoacetohydrazide | |
| 8820 | |
| ≥97.0% (T) |
| 140-87-4 | |
| C3H5N3O | |
| 25 g | |
| cyanoacetohydrazide, cyacetacide, cyanoacethydrazide, cyanoacetic acid hydrazide, cyacetacid, cyacetazid, cyacetazide, mackreazid, neohydrazid, armazal | |
| C(C#N)C(=O)NN | |
| 99.093 | |
| 99.09 | |
| Crystalline Powder |
Chemical Identifiers
| 140-87-4 | |
| 99.093 | |
| HPHBOJANXDKUQD-UHFFFAOYSA-N | |
| 8820 | |
| C(C#N)C(=O)NN |
| C3H5N3O | |
| MFCD00007611 | |
| cyanoacetohydrazide, cyacetacide, cyanoacethydrazide, cyanoacetic acid hydrazide, cyacetacid, cyacetazid, cyacetazide, mackreazid, neohydrazid, armazal | |
| 2-cyanoacetohydrazide |
Safety and Handling
RTECSNumber : AG4200000
TSCA : No