accessibility menu, dialog, popup

UserName

CAS RN 140-87-4

H2NNHON

CAS RN 140-87-4

IUPAC Name: 2-cyanoacetohydrazide
Synonyms: cyacetacide
Molecular Weight (g/mol): 99.09
Molecular Formula: C3H5N3O
InChi Key: HPHBOJANXDKUQD-UHFFFAOYSA-N
SMILES: NNC(=O)CC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results