missing translation for 'onlineSavingsMsg'
Learn More

Cyanidin chloride, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014471
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014471 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014471 Supplier Merck Emd Millipore Supplier No. 9409910MG
Food and Beverages

Specifications

CAS 528-58-5
Physical Form Neat
Linear Formula C15H11 ClO6
MDL Number MFCD00017582
Synonym 3,3 ′,4,5,7-Pentahydroxyflavylium chloride; Cyanidol chloride
Type Analytical Standard
Quantity 10 mg
Formula Weight 322.70
Percent Purity ≥98% (HPLC)
Grade Analytical Standard