missing translation for 'onlineSavingsMsg'
Learn More

Cyanide, standard solution, Specpure™ CN∣- 1000μg/mL

KCN in 0.1% NaOH

$247.22 - $483.99

Chemical Identifiers

Molecular Formula KCN in 0.1% NaOH
MDL Number MFCD00468042
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43804AE
View Documents
Thermo Scientific Chemicals
043804AE
100 mL
Each for $247.22
Only null left
 
AA43804AP
View Documents
Thermo Scientific Chemicals
043804AP
500 mL
Each for $483.99
Only null left
 
Description

Description

Cyanide, standard solution is used to dissolve metals and their ores. It is also used in clinical chemistry and in waste water treatment facilities to determine the concentration of dissolved and particulate materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

KCN in 0.1% NaOH
MFCD00468042

Specifications

Odorless
Liquid
MFCD00468042
Miscible with water
CN: 1000μg/mL
1g/cm3 at 20°C
KCN in 0.1% NaOH
Colorless
100 mL
Cyanide, Standard Solution
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Wear suitable protective clothing and gloves.

P264b-P270-P280-P301+P310-P302+P352-P312-P330-P363-P501c

H301-H312

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only