missing translation for 'onlineSavingsMsg'
Learn More

Cyanide Standard, 1000 ppm CN, Reagents

Catalog No. 01335726
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335726 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335726 Supplier Labchem Inc Supplier No. CC000400500B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cyanide Standard, 1000 ppm CN, Reagent, use for Cyanide analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Cyanide Standard, 1000 ppm CN
CAS 7732-18-5, 151-50-8, 151-50-8
Specific Gravity 1.0016
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Type Conductivity Standard
Industry Type Cyanide Analysis
Quantity 500 mL
Grade Reagent
Show More Show Less