missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cy5.5 NHS Ester Mono-Reactive CyDye

Catalog No. 45001258
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001258 5 mg
01183293 25 mg
45001195 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45001258 Supplier Cytiva Supplier No. PA15605
Only null left

Use for highly accurate labeling of amine groups. Cytiva Cy5.5 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

CyDye fluors are versatile fluorophores designed for a broad range of applications. CyDye fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye flours enables multiplexing without crosstalk for high accuracy.

Cy5.5 NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding

Specifications

Format Powdered
For Use With (Application) For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP)
Quantity 5 mg
Product Type Cy5.5 NHS Ester
Kit Contents Foil packs of premeasured dye
pH Range 3 to 10
Content And Storage 2°C to 8°C
Label or Dye Cy5.5
Color Blue