missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cy-vBRIDP(regR) 90.0+%, TCI America™
Specifications
Specifications
| Melting Point | 130°C |
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 390.55 |
| Percent Purity | ≥90.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cy-vBRIDP(regR) |
Safety and Handling
| TSCA | No |