Learn More
CXCR3 Rabbit anti-Human, Unconjugated, Clone: ACFD-3, Abnova™
Rabbit Monoclonal Antibody.
Supplier:  Abnova Corporation MAB22014
Catalog No. 89339752
Only null left
Explore available promotions
Description
Description
CXCR3 Monoclonal antibody specifically detects CXCR3 in Human samples. It is validated for WB, IP, ICC, IF.
Specifications
Specifications
CXCR3
Monoclonal
Unconjugated
CXCR3
Rabbit
100 μg
2833
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
IgG
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence
ACFD-3
chemokine (C-X-C motif) receptor 3
CD182, CD183, CKR-L2, CMKAR3, GPR9, IP10-R, Mig-R, MigR
A synthetic peptide corresponding to human CXCR3
Primary
Human
Liquid
Product Suggestions
Product Suggestions