missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CXCL15 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5150135

Catalog No. PA5150135


Only null left

Description

Description

CXCL15 Polyclonal Antibody for Western Blot, IHC, ICC/IF

Chemotactic for neutrophils. Involved in lung-specific neutrophil trafficking during normal and inflammatory conditions.
Specifications

Chemical Identifiers

CXCL15
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q9WVL7
20309
Recombinant Chemokine C-X-C-Motif Ligand 15 (CXCL15) (Gln26-Ala167).
Antibody
0.5 mg/mL
Immunohistochemistry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Mouse
Cxcl15
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 15; CXC; CXC motif chemokine; C-X-C motif chemokine 15; CXCL; Cxcl15; Il8; interleukin 8; lungkine; Scyb15; small inducible cytokine subfamily B, member 15; small-inducible cytokine B15; weche
Cxcl15
Primary
Protein A

Specifications

CXCL15
Polyclonal
Unconjugated
Cxcl15
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 15; CXC; CXC motif chemokine; C-X-C motif chemokine 15; CXCL; Cxcl15; Il8; interleukin 8; lungkine; Scyb15; small inducible cytokine subfamily B, member 15; small-inducible cytokine B15; weche
Rabbit
Protein A
RUO
20309
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q9WVL7
Cxcl15
Recombinant Chemokine C-X-C-Motif Ligand 15 (CXCL15) (Gln26-Ala167).
200 μL
Primary
Mouse
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions