Learn More
DuPont™ ProShield™ 10 Hooded Coverall
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier:  DuPont™ PB127SBU7X002500
 View more versions of this product
Catalog No. 19-162-652
Only null left
Explore available promotions
Description
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
General Purpose Coveralls
Unisex
Blue
Elastic
Hooded
Front Zipper
Non-sterile
Yes
General Maintenance, Janitorial, Cleaning
7X-Large
ProShield
Single-use
Elastic
Attached
Serged
Particle
PB127SWH
25/Cs.
Product Suggestions
Product Suggestions