missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 10 Hooded Coverall

Catalog No. p-4596739
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
7X-Large
Color:
Blue
White
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19162549 Medium Blue
19162550 Large Blue
19162551 X-Large Blue
19162552 2X-Large Blue
19162652 7X-Large Blue
19162542 Medium White
19162543 Large White
19162544 X-Large White
19162545 2X-Large White
19162546 3X-Large White
19162548 5X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19162549 Supplier DuPont™ Supplier No. PB127SBUMD002500
Only null left

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material ProShield
Color Blue
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Latex Free Yes
Model PB127SWH
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less