missing translation for 'onlineSavingsMsg'
Learn More

Curcumin (Synthetic) 97.0+%, TCI America™

Catalog No. p-7125393
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23025G 5 g
C230225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23025G Supplier TCI America Supplier No. C23025G
Only null left

Chemical Identifiers

CAS 458-37-7
Molecular Formula C21H20O6
Molecular Weight (g/mol) 368.39
MDL Number MFCD00008365
InChI Key ZIUSSTSXXLLKKK-KOBPDPAPSA-N
Synonym curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
PubChem CID 969516
ChEBI CHEBI:3962
IUPAC Name (1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
SMILES COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O

Specifications

Melting Point 186°C
Quantity 5 g
Formula Weight 368.39
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Curcumin (Synthetic)
RTECSNumber MI5230000
TSCA Yes
Recommended Storage Freezer