missing translation for 'onlineSavingsMsg'
Learn More

Curcumin (Natural), TCI America™

Supplier:  TCI America C04341G

 View more versions of this product

Catalog No. C04341G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Curcumin (Natural)
183°C
MFCD00008365
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
368.39
CHEBI:3962
Crystalline Powder
458-37-7
C21H20O6
1 g
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
(1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
969516
368.39

Chemical Identifiers

458-37-7
368.39
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
969516
(1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
C21H20O6
MFCD00008365
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
CHEBI:3962
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MI5230000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions