missing translation for 'onlineSavingsMsg'
Learn More

Cupriethylenediamine, Certified, 1.00M ±0.02M, LabChem™

Catalog No. p-5388123
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
LC135432 1 L
LC135434 4 L
2 options

Catalog No. LC135432

Supplier: LabChem LC135432

Only null left

Chemical Identifiers

CAS 13426-91-0 , 7732-18-5
Molecular Formula C4H12CuN4-2
Molecular Weight (g/mol) 179.714
InChI Key GOYYUYNOGNSLTE-UHFFFAOYSA-N
Synonym copper-ethylenediamine complex, cupriethylene diamine, bis 1,2-ethanediamine-n,n' copper 2+, koplex aquatic herbicide, unii-nip4i4lvcc, cupriethylenediamine, bis ethylenediamine copper ion, bis ethylenediamine copper 2+, bis ethylenediamine copper 2+ ion, ethane, 1,2-diamino-, copper complex
PubChem CID 4578819
IUPAC Name copper;2-azanidylethylazanide
SMILES C(C[NH-])[NH-].C(C[NH-])[NH-].[Cu+2]

Specifications

Color Blue
CAS Max % 77.5, 22.5
Physical Form Liquid
Quantity 1 L
Linear Formula C4H16CuN4
UN Number UN1761
Solubility Information Soluble in water
Formula Weight 183.74
Grade Certified
Concentration or Composition (by Analyte or Components) 1.00M ±0.02M
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.1g/mL
Decomposition Information Carbon monoxide; Carbon dioxide; Copper
Density 1.1g/mL
Chemical Name or Material Cupriethylenediamine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Refrigerate