missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate TS, 12.5% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 24944

 View more versions of this product

Catalog No. 2494-4


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cupric Sulfate
Test Solution
87.0,10.9
CuH10O9S
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
249.68
CHEBI:31440
Laboratory
Aqueous Solution
120 mL
12.5% (w/v)
Blue
7758-99-8,7732-18-5
91.0,11.34
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
24463
12.5%
Natural Poly Bottle
∼4
Liquid

Chemical Identifiers

7758-99-8
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
copper(2+) pentahydrate sulfate
CuH10O9S
MFCD00149681
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions