missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate, Anhydrous, Powder, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C141825GM

 View more versions of this product

Catalog No. 18610030


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7758-98-7
25 g
Reagent
Poly Bottle
1
98%
0.01

Chemical Identifiers

7758-98-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.