missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate, Anhydrous, Powder, 98%, Spectrum™ Chemical

Catalog No. 18610029
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610029 5 g Amber Glass Bottle
18610030 25 g Poly Bottle
18610031 125 g Poly Bottle
18610032 500 g Poly Bottle
18610033 2.5 kg Poly Bottle
18610034 12 kg Poly Pail
18610035 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18610029 Supplier Spectrum Chemical Mfg Cor Supplier No. C14185GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7758-98-7

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 98%
Grade Reagent
Loss on Drying 0.01
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.