missing translation for 'onlineSavingsMsg'
Learn More

Cupric Oxide, Black, Powdered, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165361
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610020 25 g Amber Glass Bottle
18610021 125 g Amber Glass Bottle
18610022 500 g Amber Glass Bottle
18610023 2.5 kg Amber Glass Bottle
18610024 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610020 Supplier Spectrum Chemical Mfg Cor Supplier No. C141525GM
Only null left

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.