missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Nitrate, Hydrate, Crystal, Purified, Spectrum™ Chemical
C1407, 19004-19-4, Cu(NO3)2·2.5H2O
$277.46 - $11014.18
Chemical Identifiers
| CAS | 19004-19-4 |
|---|---|
| Molecular Formula | Cu2H10N4O17 |
| Molecular Weight (g/mol) | 465.18 |
| InChI Key | IZDJJEMZQZQQQQ-UHFFFAOYSA-N |
| IUPAC Name | dicopper(2+) pentahydrate tetranitrate |
| SMILES | O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601907
|
Spectrum Chemical Mfg Cor
C1407125GM |
125 g | Poly Bottle |
Each for $277.46
|
|
||||
|
18601908
|
Spectrum Chemical Mfg Cor
C1407500GM |
500 g | Poly Bottle |
Each for $702.04
|
|
||||
|
18601909
|
Spectrum Chemical Mfg Cor
C140725KG |
2.5 kg | Poly Pail |
Each for $2,547.25
|
|
||||
|
18601910
|
Spectrum Chemical Mfg Cor
C140712KG |
12 kg | Poly Pail |
Each for $5,278.36
|
|
||||
|
18601911
|
Spectrum Chemical Mfg Cor
C140745KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Cupric Nitrate, Hydrate, Crystal, Purified is a also known as a copper nitrate and can be obtained as a crystalline solid as well as a hydrated form. Mostly used to produce copper oxide, it is also used as a polisher for other metals and in pyrotechnics.Chemical Identifiers
| 19004-19-4 | |
| 465.18 | |
| dicopper(2+) pentahydrate tetranitrate |
| Cu2H10N4O17 | |
| IZDJJEMZQZQQQQ-UHFFFAOYSA-N | |
| O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| 19004-19-4 | |
| Cu2H10N4O17 | |
| 125 g | |
| O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 465.18 | |
| Poly Bottle |
| 100% | |
| UN1477 | |
| IZDJJEMZQZQQQQ-UHFFFAOYSA-N | |
| dicopper(2+) pentahydrate tetranitrate | |
| Purified |