missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate, Hydrate, Crystal, Purified, Spectrum™ Chemical

Catalog No. 18601908
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
125 g
2.5 kg
45 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18601908 500 g Poly Bottle
18601907 125 g Poly Bottle
18601909 2.5 kg Poly Pail
18601910 12 kg Poly Pail
18601911 45 kg Fiber Drum
5 options

Catalog No. 18601908

Supplier: Spectrum Chemical Mfg Cor C1407500GM

Only null left

C1407, 19004-19-4, Cu(NO3)2·2.5H2O

Spectrum™ Chemical Cupric Nitrate, Hydrate, Crystal, Purified is a also known as a copper nitrate and can be obtained as a crystalline solid as well as a hydrated form. Mostly used to produce copper oxide, it is also used as a polisher for other metals and in pyrotechnics.

Chemical Identifiers

CAS 19004-19-4
Molecular Formula Cu2H10N4O17
Molecular Weight (g/mol) 465.18
InChI Key IZDJJEMZQZQQQQ-UHFFFAOYSA-N
IUPAC Name dicopper(2+) pentahydrate tetranitrate
SMILES O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Max % 100%
UN Number UN1477
Quantity 500 g
Grade Purified
Packaging Poly Bottle