missing translation for 'onlineSavingsMsg'
Learn More

Cupric Bromide, Crystal, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C137550KGBL

 View more versions of this product

Catalog No. 18609977

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

7789-45-9
50 kg
Reagent
1
98%
Fiber Drum

Chemical Identifiers

7789-45-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.