missing translation for 'onlineSavingsMsg'
Learn More

Cupric Acetate, Monohydrate, Purified, Spectrum™ Chemical

$526.25 - $4458.82

Chemical Identifiers

CAS 6046-93-1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609971
View Documents
Spectrum Chemical Mfg Cor
C1371500GM
500 g Poly Bottle
Each for $526.25
Only null left
 
18609972
View Documents
Spectrum Chemical Mfg Cor
C137125KG
2.5 kg Poly Pail
Each for $1,810.25
Only null left
 
18609973
View Documents
Spectrum Chemical Mfg Cor
C137112KG
12 kg Poly Pail
Each for $4,458.82
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6046-93-1

Specifications

6046-93-1
500 g
Poly Bottle
1
Purified
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.