missing translation for 'onlineSavingsMsg'
Learn More

Cupric Acetate, Monohydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18609966
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609966 125 g Poly Bottle
18609967 500 g Poly Bottle
18609968 2.5 kg Poly Bottle
18609969 12 kg Poly Pail
18609970 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609966 Supplier Spectrum Chemical Mfg Cor Supplier No. C1370125GM

Chemical Identifiers

CAS 6046-93-1

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98 to 102%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.