missing translation for 'onlineSavingsMsg'
Learn More

Cupferron 98.0+%, TCI America™

Catalog No. C064625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C064625G 25 g
C0646500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C064625G Supplier TCI America Supplier No. C064625G

Chemical Identifiers

CAS 135-20-6
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00078422
InChI Key NJVHCUNZAMFQNA-UHFFFAOYSA-N
Synonym cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
PubChem CID 2724103
IUPAC Name N-nitroso-N-phenylhydroxylamine amine
SMILES N.ON(N=O)C1=CC=CC=C1

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 155.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cupferron
EINECSNumber (9)-2320&(3)-2750
RTECSNumber NC4725000
TSCA Yes
Recommended Storage Refrigerator