missing translation for 'onlineSavingsMsg'
Learn More

Cupferron, 97+%

Catalog No. AAA1655114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1655114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1655114 Supplier Thermo Scientific Chemicals Supplier No. A1655114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135-20-6 | C6H9N3O2 | 155.16 g/mol

Reagent for determination of Ce, Cu, Fe, Sn, and Ti

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135-20-6
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00078422
InChI Key NJVHCUNZAMFQNA-UHFFFAOYSA-N
Synonym cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
PubChem CID 2724103
IUPAC Name azanium;N-oxido-N-phenylnitrous amide
SMILES N.ON(N=O)C1=CC=CC=C1

Specifications

Melting Point 154°C (decomposition)
Odor Odorless
Quantity 25 g
UN Number UN2811
Merck Index 14,2622
Formula Weight 155.16
Percent Purity ≥97%
Chemical Name or Material Cupferron
Hazard Category H301-H315-H319-H335-H351
Hazard Statement GHS H Statement
H301-H351-H315-H319-H317
Toxic if swallowed.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 205-183-2
RTECSNumber NC4725000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only