missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cuminaldehyde 97.0+%, TCI America™
$49.36 - $499.34
Chemical Identifiers
| CAS | 122-03-2 |
|---|---|
| Molecular Formula | C10H12O |
| Molecular Weight (g/mol) | 148.21 |
| MDL Number | MFCD00006953 |
| InChI Key | WTWBUQJHJGUZCY-UHFFFAOYSA-N |
| Synonym | 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl |
| PubChem CID | 326 |
| ChEBI | CHEBI:28671 |
| IUPAC Name | 4-(propan-2-yl)benzaldehyde |
| SMILES | CC(C)C1=CC=C(C=O)C=C1 |
Chemical Identifiers
| C10H12O | |
| MFCD00006953 | |
| 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl | |
| CHEBI:28671 | |
| CC(C)C1=CC=C(C=O)C=C1 |
Specifications
| 122-03-2 | |
| 240°C | |
| MFCD00006953 | |
| WTWBUQJHJGUZCY-UHFFFAOYSA-N | |
| 4-(propan-2-yl)benzaldehyde | |
| 326 | |
| 148.21 | |
| Liquid |
| Yellow | |
| C10H12O | |
| 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl | |
| CC(C)C1=CC=C(C=O)C=C1 | |
| 148.21 | |
| CHEBI:28671 | |
| ≥97.0% (GC) | |
| Cuminaldehyde |
Safety and Handling
EINECSNumber : (3)-1144&(3)-2663
RTECSNumber : CU7000000
TSCA : Yes