missing translation for 'onlineSavingsMsg'
Learn More

Cuminaldehyde 97.0+%, TCI America™

Catalog No. I016825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016825G 25 g
I0168500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I016825G Supplier TCI America Supplier No. I016825G

Chemical Identifiers

CAS 122-03-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00006953
InChI Key WTWBUQJHJGUZCY-UHFFFAOYSA-N
Synonym 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
PubChem CID 326
ChEBI CHEBI:28671
IUPAC Name 4-(propan-2-yl)benzaldehyde
SMILES CC(C)C1=CC=C(C=O)C=C1

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 g
Formula Weight 148.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cuminaldehyde
EINECSNumber (3)-1144&(3)-2663
RTECSNumber CU7000000
TSCA Yes