missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cuminaldehyde 97.0+%, TCI America™
Supplier: TCI America I0168500G
Specifications
| Cuminaldehyde | |
| Yellow | |
| C10H12O | |
| 500 g | |
| WTWBUQJHJGUZCY-UHFFFAOYSA-N | |
| 4-(propan-2-yl)benzaldehyde | |
| 326 | |
| 148.21 | |
| Liquid |
| 122-03-2 | |
| 240°C | |
| MFCD00006953 | |
| 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl | |
| CC(C)C1=CC=C(C=O)C=C1 | |
| 148.21 | |
| CHEBI:28671 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 122-03-2 | |
| 148.21 | |
| WTWBUQJHJGUZCY-UHFFFAOYSA-N | |
| 326 | |
| 4-(propan-2-yl)benzaldehyde |
| C10H12O | |
| MFCD00006953 | |
| 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl | |
| CHEBI:28671 | |
| CC(C)C1=CC=C(C=O)C=C1 |
Safety and Handling
EINECSNumber : (3)-1144&(3)-2663
RTECSNumber : CU7000000
TSCA : Yes