missing translation for 'onlineSavingsMsg'
Learn More

Cumene 99.0+%, TCI America™

Catalog No. C0687500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0687500ML 500 mL
C068725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0687500ML Supplier TCI America Supplier No. C0687500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008881
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
Synonym isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1

Specifications

Melting Point -96°C
Color Colorless
Boiling Point 155°C
Quantity 500 mL
UN Number 1918
Formula Weight 120.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cumene
EINECSNumber (3)-0022
RTECSNumber GR8575000
TSCA Yes