missing translation for 'onlineSavingsMsg'
Learn More

Cumene Hydroperoxide (contains ca. 20% Aromatic Hydrocarbon), TCI America™

Catalog No. C2223100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2223100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2223100G Supplier TCI America Supplier No. C2223100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80-15-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002129
InChI Key YQHLDYVWEZKEOX-UHFFFAOYSA-N
Synonym cumene hydroperoxide, cumyl hydroperoxide, hydroperoxide, 1-methyl-1-phenylethyl, cumenyl hydroperoxide, alpha,alpha-dimethylbenzyl hydroperoxide, cumolhydroperoxid, 7-cumyl hydroperoxide, cumolhydroperoxide, cument hydroperoxide, hydroperoxyde de cumene
PubChem CID 6629
ChEBI CHEBI:78673
IUPAC Name 2-phenylpropane-2-peroxol
SMILES CC(C)(OO)C1=CC=CC=C1

Specifications

Melting Point -37°C
Color Yellow
Quantity 100 g
UN Number 3109
Formula Weight 152.19
Physical Form Liquid
Chemical Name or Material Cumene Hydroperoxide (contains ca. 20% Aromatic Hydrocarbon)
EINECSNumber (3)-1014
RTECSNumber MX2450000
TSCA Yes