Learn More
Cumene, 99.9%, extra pure
CAS: 98-82-8 | C9H12 | 120.19 g/mol
Supplier: Thermo Scientific Chemicals 329730025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cumene | |
| 98-82-8 | |
| Colorless | |
| 152.0°C to 154.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.4905 to 1.4925 | |
| MFCD00008881 | |
| 1918 | |
| 0.862 | |
| isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano | |
| RWGFKTVRMDUZSP-UHFFFAOYSA-N | |
| cumene | |
| 7406 | |
| CHEBI:34656 | |
| 99.9% | |
| Liquid |
| 99.9% | |
| -96.0°C | |
| 0.8620g/mL | |
| 31°C | |
| 99.85% min. (GC) | |
| C9H12 | |
| C6H5CH(CH3)2 | |
| 2.5 L | |
| 05,393 | |
| 15,2607 | |
| Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol and many other organic solvents,carbon tetrachloride,ether,benzene,acetone | |
| CC(C)C1=CC=CC=C1 | |
| 120.19 | |
| 0.79 mPa.s (20°C) | |
| 120.19 | |
| Extra Pure |
Chemical Identifiers
| 98-82-8 | |
| 120.19 | |
| RWGFKTVRMDUZSP-UHFFFAOYSA-N | |
| 7406 | |
| cumene |
| C9H12 | |
| MFCD00008881 | |
| isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano | |
| CHEBI:34656 | |
| CC(C)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortabl
GHS Signal Word: Danger
EINECSNumber : 202-704-5
RTECSNumber : GR8575000
TSCA : TSCA
RUO – Research Use Only