missing translation for 'onlineSavingsMsg'
Learn More

Cumene, 99%

CAS: 98-82-8 | C9H12 | 120.195 g/mol

$43.78 - $195.95

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008881
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
Synonym isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA11864AK
View Documents
Thermo Scientific Chemicals
A11864AK
250 mL
Each for $43.78
Only null left
 
AAA11864AP
View Documents
Thermo Scientific Chemicals
A11864AP
500 mL
Each for $56.33
Only null left
 
AAA118640F
View Documents
Thermo Scientific Chemicals
A118640F
2500 mL
Each for $195.95
Only null left
 
Description

Description

Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

Solubility
Soluble in alcohol, ether, acetone, benzene, carbon tetrachloride. Insoluble in water.

Notes
Store at 2-8°C. Avoid prolonged storage with exposure to air. Incompatible with acids, oxidizing materials.
Specifications

Chemical Identifiers

98-82-8
120.195
RWGFKTVRMDUZSP-UHFFFAOYSA-N
7406
cumene
C9H12
MFCD00008881
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
CHEBI:34656
CC(C)C1=CC=CC=C1

Specifications

98-82-8
0.864
46°C (114°F)
C9H12
MFCD00008881
UN1918
14,2617
Soluble in alcohol,ether,acetone,benzene,carbon tetrachloride. Insoluble in water.
CC(C)C1=CC=CC=C1
120.195
CHEBI:34656
99%
Cumene
-96°C
152°C to 154°C
99%
1.491
250 mL
1236613
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
RWGFKTVRMDUZSP-UHFFFAOYSA-N
cumene
7406
120.2
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226-H302-H335

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P271-P281-P301+P310-P303+P361+P353-P304+P340-P308+P313-P331-P370+P378q-P501c

H226-H304-H335-H350

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOPROPYLBENZENE

EINECSNumber : 202-704-5

RTECSNumber : GR8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only